Learn More
Medchemexpress LLC Hdac-in-55 | 1268674-16-3 | 98.4% | 311.34 g/mol | C17H17N3O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15360710MG
Encompass_Preferred
HDAC-IN-55 is a small-molecule histone deacetylase (HDAC) inhibitor (CAS 1268674-16-3) reported to increase E-cadherin expression and inhibit cancer cell proliferation. It is provided as a research-grade compound for in vitro and preclinical studies; molecular weight 311.34 g/mol and chemical formula C17H17N3O3.
  • Increases E-cadherin expression and inhibits cancer cell proliferation in reported studies.
  • Research-grade purity (98.4%) suitable for biochemical and cellular assays.
  • High solubility in DMSO (100 mg/mL); ultrasonic agitation recommended.
  • Stable as a powder under low-temperature storage (powder: -20°C for long-term).
  • Intended for in vitro and preclinical research use only.
Catalog No. 50-002-86346