Learn More
TARGETMOL CHEMICALS INC HDAC-IN-57 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T773345MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. HDAC-IN-57 is an orally active pan-inhibitor of histone deacetylase (HDAC) inhibiting HDAC1 HDAC2 HDAC6 and HDAC8 with IC50 values of 2.07 nM 4.71 nM 2.4 nM and 107 nM respectively. HDAC-IN-57 inhibits LSD1 with an IC50 value of 1.34 uu. HDAC-IN-57 has anti-tumor activity and can induce apoptosis. purity: 98%
Catalog No. 50-257-9154