Learn More
Enzo Life Sciences HDAC substrate (fluorogenic) (1mg). CAS: 233691-67-3
SDP
Supplier:  Enzo Life Sciences ALX260137M001
Encompass_Preferred
A fluorogenic substrate for histone deacetylase (HDAC). Ex(max): 390nm; Em(max): 460nm. Alternative name: Boc-Lys(Ac)-AMC, MAL, Histone deacetylase substrate (fluorogenic), N-(4-Methyl-7-aminocoumarinyl)-Nalpha-(t-butoxycarbonyl)-Nomega-acetyllysineamide. Formula: C23H31N3O6. MW: 445.5. Purity: ≥99%. Appearance: White crystalline solid. Solubility: Soluble in DMSO and 100% ethanol (1mg/ml). Long Term Storage: -20°C. Handling: Protect from light.
Catalog No. 50-200-8340