Learn More
Enzo Life Sciences FLUOR DE LYS HDAC1 fluorometric drug discovery assay kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences BMLAK5110001
Encompass_Preferred
A FLUOR DE LYS fluorescent assay system. The HDAC1 Fluorescent Activity Assay/Drug Discovery Kit is a complete assay system designed to measure the lysyl deacetylase activity of the recombinant human HDAC1 included in the kit. The kit is ideal for chemical library screening for candidate inhibitors or kinetic assay of the enzyme under varying conditions. The FLUOR DE LYS HDAC1 assay is based on the FLUOR DE LYS Substrate and FLUOR DE LYS Developer combination. The assay procedure has two steps. First, the FLUOR DE LYS Substrate, which comprises an acetylated lysine side chain, is incubated with HDAC1. Deacetylation of the substrate sensitizes the substrate so that, in the second step, treatment with the FLUOR DE LYSDeveloper produces a fluorophore. Alternative name: Histone deacetylase 1 fluorescent assay kit. Applications: Fluorescent detection, HTS Cell-based assays, Activity assay.
Catalog No. 50-201-0285