Learn More
Medchemexpress LLC HDAC10-IN-2 (hydrochloride) | 95.0% | C19H23ClN2O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY144782A
Encompass_Preferred
HDAC10-IN-2 hydrochloride is a potent and highly selective HDAC10 inhibitor with an IC50 of 20 nM. This compound modulates autophagy in aggressive FLT3-ITD positive acute myeloid leukemia cells and is for research use only.
  • Potent and highly selective HDAC10 inhibitor
  • IC50 of 20 nM (drHDAC10)
  • Modulates autophagy in aggressive FLT3-ITD positive acute myeloid leukemia cells
  • Relevant to epigenetics and autophagy pathways
Catalog No. 50-003-53527