Learn More
Medchemexpress LLC Hdac10-in-2 hydrochloride | 95.0% | C19H23ClN2O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY144782A50MG
Encompass_Preferred
HDAC10-IN-2 hydrochloride is a potent and highly selective HDAC10 inhibitor with an IC50 of 20 nM. It modulates autophagy in aggressive FLT3-ITD positive acute myeloid leukemia cells.
  • Potent and highly selective HDAC10 inhibitor.
  • IC50 of 20 nM for drHDAC10.
  • Modulates autophagy in aggressive FLT3-ITD positive acute myeloid leukemia cells.
  • For research use only.
Catalog No. 50-003-54436