Learn More
Sino Biological Recombinant Human HDAC7 Protein (GST Tag), Active
Supplier:  Sino Biological H8931G50
Encompass_Preferred
Recombinant human HDAC7 (501-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4691