Learn More
Sino Biological Recombinant Human HDAC1 Protein (Full Length, GST Tag), Active
Supplier:  Sino Biological H8330G500
Encompass_Preferred
Recombinant full length human HDAC1 was expressed by baculovirus in Sf9 insect cells using a C-terminal GST tag.
Catalog No. 50-257-4714