Learn More
Cayman Chemical HDAC6 InhIbItr 1mg
SDP
Supplier:  Cayman Chemical 152001 MG
Encompass_Preferred
A potent and selective inhibitor of HDAC6 (IC50 = 36 nM) that poorly blocks other HDAC enzymes; cell permeable, inhibiting the acetylation of tubulin in cells with an IC50 value of 210 nM
Catalog No. 50-297-2951