Learn More
Cayman Chemical HDAC6 InhIbItr 500ug
SDP
Supplier:  Cayman Chemical 15200500 UG
Encompass_Preferred
A potent and selective inhibitor of HDAC6 (IC50 = 36 nM) that poorly blocks other HDAC enzymes; cell permeable, inhibiting the acetylation of tubulin in cells with an IC50 value of 210 nM
Catalog No. 50-296-9180