Learn More
Cayman Chemical HDAC7humnrecombInt 10ug
SDP
Supplier:  Cayman Chemical 4208410 UG
Encompass_Preferred
Source: Active recombinant human N-terminal GST-tagged HDAC7 expressed in insect cells • Amino acids: 518-952 • MW: 78 kDa
Catalog No. 50-294-5718