Learn More
Enzo Life Sciences HDAC8 (human), (recombinant) (100 U)
SDP
Supplier:  Enzo Life Sciences BMLSE1450100
Encompass_Preferred
Alternative name: Histone deacetylase 8. Formulation: Liquid. In 10mM TRIS, pH 7.5, containing 100mM NaCl, 3mM MgCl2 and 10% glycerol. Source: Produced in E. coli. Recombinant HDAC8 from human cDNA (377 aa). Long Term Storage: -80°C.
Catalog No. 50-201-1941