Learn More
Medchemexpress LLC HDAC8-IN-1 | 1417997-93-3 | 99.8% | 345.39 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY111342
Encompass_Preferred
HDAC8-IN-1 is a highly selective inhibitor of Histone Deacetylase 8 (HDAC8) with an IC50 of 27.2 nM. It demonstrates antiproliferative effects in human lung cancer cell lines such as A549, H1299, and CL1-5, and exhibits cytotoxicity against human lung CL1-5 cells without significant cytotoxicity towards normal IMR-90 cells. This compound has also been observed to slightly reverse IL-4-induced Foxp3 repression, acting similarly to a class I HDAC inhibitor.
  • Potent inhibitor with an IC50 of 27.2 nM.
  • Shows antiproliferative effects in various human lung cancer cell lines.
  • Exhibits selective cytotoxicity against human lung CL1-5 cells, with minimal impact on normal IMR-90 cells.
  • Can slightly reverse IL-4-induced Foxp3 repression and promote regulatory T cell differentiation.
Catalog No. 50-003-46817