Learn More
Medchemexpress LLC hDHODH-IN-13 | 3032611-13-2 | 100.0% | 393.84 g·mol⁻¹ | C21H17ClFN5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY14964210MG
Encompass_Preferred
hDHODH-IN-13 is a small-molecule inhibitor of human dihydroorotate dehydrogenase (hDHODH) used in research applications such as inflammatory bowel disease (IBD) models and DHODH biology studies. The compound shows an IC50 of 173.4 nM against hDHODH, has molecular formula C21H17ClFN5, molecular weight 393.84 g·mol⁻¹, and CAS number 3032611-13-2. It is supplied with high stated purity and is available in small research-scale pack sizes suitable for biochemical and cell-based assays.
  • Inhibits human dihydroorotate dehydrogenase with low-nanomolar potency.
  • Suitable for biochemical and cell-based assays.
  • Supplied with high stated purity (99.97%).
  • Available in multiple small-pack sizes, including 10 mg.
  • Useful for research into inflammatory bowel disease and DHODH biology.
Catalog No. 50-002-87527