Learn More
TARGETMOL CHEMICALS INC HDHODH-IN-5 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T110295MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. hDHODH-IN-5 is an inhibitor of human dihydroorotate dehydrogenase (hDHODH IC50 = 0.91 uM). purity: 98%
Catalog No. 50-258-1590