Learn More
SEKISUI DIAGNOSTICS LLC HDL CHOL VERIFIER KIT
Supplier:  SEKISUI DIAGNOSTICS LLC 9560
Encompass_Preferred
HDL Cholesterol Verifier Kit is a liquid, in vitro diagnostic product intended to be used for the calibration verification of serum high density (HDL) and low density (LDL) lipoprotein cholesterol test systems.
Catalog No. NC9075284