Learn More
Selleck Chemical LLC HDM201 (Siremadlin) 1mg 1448867-41-1 NVP-HDM201
SDP
Supplier:  Selleck Chemical LLC S86061MG
Encompass_Preferred
HDM201 (Siremadlin, NVP-HDM201) is a novel, highly potent and selective inhibitor of the p53-Mdm2 interaction with affinity constant for Mdm2 in the picomolar range and a selectivity ratio greater than 10000-fold vs Mdm4. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8130