Learn More
Grainger HDMI Port Saver 8Inch Black
Supplier:  Grainger 14C362
Encompass_Preferred
The Monoprice HDMI Adapter is a passive adapter that converts an HDMI signal to DVI. It supports resolutions up to 1920x1200 and 1080p. The adapter has 24k gold-plated connectors and bare copper conductors for optimal signal quality. It features a compact design with molded connectors and no external power required. The adapter works in one direction only, converting HDMI to DVI, and does not transmit audio signals. It comes with a lifetime warranty from Monoprice.
Catalog No. 50-274-8960