Learn More
Ace Glass, Inc. Head, 4in conical flange, four neck, 45/50 center, (2)24/40, 14/35
SDP
Supplier:  Ace Glass, Inc. 648840
Encompass_Preferred
4 neck reaction flask head with 4(100mm) conical flange. Has 45/50 reinforced outer center neck, and two 29/42 reinforced outer joints, in-line, plus a reinforced 14/35 joint for use with a bushing adapter and/or thermometer.
Catalog No. 50-978-771