Learn More
Chemglass Life Sciences Chemglass Headplate for 15 L Bioreactors, 19 Ports - CLS-1385-03
SDP
Supplier:  Chemglass Life Sciences CLS138503
Encompass
The Chemglass CLS-1385-03 headplate is a replacement headplate designed for 15 L bioreactor vessels. It is machined from a single piece of 316L stainless steel, electropolished for a mirror finish, and supplied complete with a silicone o-ring to mate to the glass vessel.
  • Weld-free, machined from a single piece of 316L stainless steel.
  • Electropolished mirror finish for corrosion resistance.
  • Supplied with a silicone o-ring to mate to the glass vessel.
  • 19 ports: 69 mm center with 4 bolt holes; 6 x M27; 2 x M12; 10 x M10.
  • Intended for use on 15 L bioreactor vessels; suitable for cell culture and microbial applications.
Catalog No. 50-273-3310