Learn More
Roboz Surgical HEART HOLDING FORCEPS
SDP
Supplier:  Roboz Surgical RS5255
Encompass
Singley Heart Holding Forceps; 6 in. Length The Singley heart holding forceps are designed to hold the heart of rats without being concerned about a sharp tip damaging the heart. A pin and screw allows you to adjust the opening of the tip to accomm
Catalog No. NC9640405