Learn More
Med Vet International Heartworm Rack, 8 3/8” x 3/8” x 4 3/8”, J0352H, Each
Supplier:  Med Vet International JOR352H
Encompass_Preferred
Commonly used for heartworm testing. Holds 72 micro-sample tubes.
Catalog No. 50-005-85437