Learn More
Chemglass Life Sciences Heat Exchanger Loop for 5 L Chemglass Bioreactors (CLS-1382-11)
SDP
Supplier:  Chemglass Life Sciences CLS138211
Encompass
The Chemglass Life Sciences CLS-1382-11 heat exchanger loop is a loop-style exchanger designed to provide temperature control for 5 L bioreactor systems. It installs into two M10 ports on the reactor headplate and is used with recirculating heat-transfer fluid to maintain process temperature.
  • Loop-style heat exchanger for recirculating heat-transfer fluid.
  • Compatible with 5 L bioreactor vessels.
  • Installs into two M10 headplate ports.
  • Uses a silicone o-ring for sealing; replacement o-rings available.
Catalog No. 50-273-3315