Learn More
Cayman Chemical Heat shck protin InhIbItr 25mg
SDP
Supplier:  Cayman Chemical 1539525 MG
Encompass_Preferred
A benzylidene lactam compound that prevents the synthesis of inducible Hsps Hsp105, Hsp72, and Hsp40; at 100 μM, inhibits the development of thermotolerance in COLO 320 DM cells
Catalog No. 50-297-1397