Learn More
V&P Scientific Inc REACTION BLOCK, Holds 48, 11.8mm Diameter Vials, Eddy Current Design
Supplier:  V&P Scientific Inc VP 416ALB48
Encompass
REACTION BLOCK, for Heating and Mixing on Tumble Stirrer, Holds 48, 11.8mm Diameter Vials, Anodized Aluminum, Eddy Current Design, SLAS Footprint
Catalog No. NC2166127