Learn More
Euthanex The HB-163 Chamber Warmer / Heated Bed for heating the Sure-Seal mouse/rat induction boxes.
Supplier:  Euthanex HB163
Encompass_Preferred
The HB-163 Chamber Warmer / Heated Bed is for use with the EZ-HEAT Surgical Bed Warming System. The temperature ranges from 85 to 104 degrees Fahrenheit. Use with the EZ-178 SURE-SEAL MOUSE/RAT CHAMBER or by itself as an extra large heated surgical bed by adding the EZ-103H Microflex Breather.
Catalog No. NC1695016