Learn More
Caron Products And Services Heated Incubator. 25 cubic foot,
Supplier:  Caron Products And Services 7002251
Encompass_Preferred
Heated Incubator. 25 cubic foot with temperature control, touchscreen. 10C above ambient to 70C, 115V, 60 Hz.
Catalog No. NC2267248
Only null left
Add to Cart
Request a Quote