Learn More
Medchemexpress LLC Heclin 10mM | 890605-54-6 | 98.2% | 283.32 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11020410MM/1ML
Encompass_Preferred
Heclin is a HECT E3 ubiquitin ligases inhibitor. It inhibits Smurf2, Nedd4, and WWP1 with IC50 values of 6.8, 6.3, and 6.9 μM, respectively, and can be used for the research of gastric cancer.
  • Functions as a HECT E3 ubiquitin ligases inhibitor.
  • Inhibits Smurf2, Nedd4, and WWP1.
  • Useful for gastric cancer research.
  • Inhibits cell viability and IGF1 signaling in BGC803 and MKN45 cell lines.
  • Ameliorates epinecidin-1-mediated MyD88 degradation in Raw264.7 cells.
Catalog No. 50-003-45116