Learn More
Medchemexpress LLC Hederacoside D | 760961-03-3 | 99.5% | 1075.24 | 25 MG

Supplier: Medchemexpress LLC 25MGHYN0254
Hederacoside D is one of the bioactive saponins from Hedera helix, playing pivotal roles in the overall biological activity. It is intended for research use only.
- Appearence: Solid
- Color: White to off-white
- Structure classification: Terpenoids, Triterpenes
- Initial source: Plants, Araliaceae, Hedera nepalensis K. Koch var. sinensis (Tobl.) Rehd.
- Solubility: In DMSO: 100 mg/mL (93.00 mM; Need ultrasonic; Hygroscopic DMSO has a significant impact on the solubility of product, please use newly opened DMSO)
- Pharmacokinetic behavior successfully applied in rats after oral and intravenous administration.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.