Learn More
Cygnus Technologies Cygnus Technologies HEK 293 Antigen Concentrate, Supports Accurate Analyte Control Preparation, Compatible With ELISA Kits, 12 Units
Supplier:  Cygnus Technologies F653SX
Encompass
The HEK 293 Antigen Concentrate by Cygnus Technologies is specifically designed for applications involving Human Embryonic Kidney (HEK) 293 host cell proteins. This specialized concentrate facilitates accurate preparation of analyte controls for ELISAs and aids in monitoring host cell impurity removal during bioprocess development and quality control. Its capability to support critical spike and recovery studies makes it a valuable tool for researchers in the biotechnology field.
Catalog No. NC2176350