Learn More
Eurofins Discoverx PathHunter HEK 293 cyno CXCR2 β-Arrestin Cell Line, 2 vials. (93-0981C1)
Supplier:  Eurofins Discoverx 930981C1
Encompass
The PathHunter cyno CXCR2 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CXCL8/IL8) based activation of CXCR2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged cyno CXCR2 and EA-tagged β-Arrestin-2. Activation of CXCR2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Cynomolgus monkey version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1737
Add to Cart
Request a Quote