Learn More
Invivogen HEK-Blue hTLR2, 3-7 x 10e6 cells
SDP
Supplier:  Invivogen HKBHTLR2
Encompass
HEK-Blue-hTLR2 cells were obtained by co-transfection of the human TLR2 and SEAP (secreted embryonic alkaline phosphatase) genes into HEK293 cells.
Catalog No. NC0420989