Learn More
Invivogen HEK-Blue hTLR4, 3-7 x 10e6 cells
SDP
Supplier:  Invivogen HKBHTLR4
Encompass
HEK-Blue-hTLR4 cells were obtained by co-transfection of the human TLR4, MD-2 and CD14 co-receptor genes, and an inducible SEAP (secreted embryonic alkaline phosphatase) reporter gene into HEK293 cells.
Catalog No. NC9040639