Learn More
Invivogen HEK-Blue™ IFN-α/β Cells, Reporter System for IFN Detection, Mycoplasma-free
SDP
Supplier:  Invivogen HKBIFNABV2
Encompass
The HEK-Blue™ IFN-α/β Cells are a specialized reporter cell line designed for the detection of bioactive human type I interferons (IFN-α and IFN-β). Engineered from HEK 293 cells, they integrate a secreted alkaline phosphatase (SEAP) reporter gene for precise cytokine bioassays. This system uniquely responds to IFN-α and IFN-β, providing a reliable colorimetric readout using QUANTI-Blue™ Solution, making it essential for researchers targeting the IFN pathway.
Catalog No. NC2683657
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.