Learn More
Genscript Corporation HEK293/EP1 Stable Cell Line
Supplier:  Genscript Corporation M00228
Encompass
Recombinant HEK293 cells stably overexpress human prostaglandin E receptor 1 (PTGER1) on the surface and contain high levels of G protein G q to couple with the receptor in downstream signaling pathways
Catalog No. 50-000-57862
Add to Cart
Request a Quote