Learn More
Genscript Corporation HEK293/G15 Stable Cell Line
Supplier:  Genscript Corporation M00554
Encompass
HEK293/G 15 is a HEK293 cell line stably expressing the G 15 alpha subunit protein which a Gq protein It is used as a host cell for transfection expression of Gs and Gi/o -coupled receptors the constitutively expressed G 15 protein in the cells allows many transfected receptors which normally stimulate/inhibit the cAMP pathway to couple to Gq signal transduction and mobilize intracellular calcium The cell line carries the puromycin resistance gene and is resistant to puromycin
Catalog No. 50-000-58057
Only null left
Add to Cart
Request a Quote