Learn More
Eurofins Discoverx InCELL Hunter HEK 293 SMARCA4 Bromodomain Cell Line, 2 vials. (96-0060C1)
Supplier:  Eurofins Discoverx 960060C1
Encompass
The InCELL Hunter HEK 293 SMARCA4 Bromodomain Cell Line is a stable, engineered cell line for use in studying small molecule drug candidates that engage with regulators of epigenetics pathways. This cell line enables assessment of compound-engagement-based stabilization of SMARCA4 via detection of SMARCA4 levels. This product uses EFC technology. The cell line included overexpresses SMARCA4 target protein fused to ProLabel. In the absence of compound that binds and stabilizes the target, this protein is degraded and no EFC signal is produced. When compound engages and stabilizes the target, EA contained in the detection reagent produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/epigenetics.
Catalog No. 50-200-4487
Add to Cart
Request a Quote