Learn More
Enzo Life Sciences HEK293T host cell protein ELISA kit
SDP
Supplier:  Enzo Life Sciences ENZKIT1620001
Encompass_Preferred
The HEK293T host cell protein ELISA kit is a complete, colorimetric, immunometric immunoassay kit for the quantitative determination of HEK293T host cell protein contamination with results in 6 hours.
Catalog No. NC1663133