Learn More
Enzo Life Sciences HeLa (heat shocked) (no recovery time), (cell lysate) (200µg)
SDP
Supplier:  Enzo Life Sciences ADILYCHL102F
Encompass_Preferred
Formulation: Liquid. In SDS-PAGE sample buffer. Source: Human. Long Term Storage: -80°C. Handling: Avoid freeze/thaw cycles. Applications: WB .
Catalog No. 50-200-7334