Learn More
Cellomics Technology LLC Hela/GFP-firefly Luciferase (CMV) Stable Cells, 2x106 Cells
SDP
Supplier:  Cellomics Technology LLC SC1273
Encompass
Hela, a human cervix adenocarcinoma cell line, has been widely used in cancer research and drug development in vitro and in vivo. Hela/GFP-firefly luciferase (CMV) stable cells express transgenically GFP-firefly luciferase under the control of the CMV promoter.Hela cell line was from human cervix adenocarcinoma. It has been widely used in cancer research and drug development. Hela/GFP-firefly luciferase (CMV) stable cells express GFP-firefly luciferase under the control of the CMV promoter. These cells can be used for in vitro cell model and in vivo small-animal imaging.
Catalog No. NC2748929