Learn More
Enzo Life Sciences HeLa (heat shocked) , (cell lysate) (200 µg)
SDP
Supplier:  Enzo Life Sciences ADILYCHL101F
Encompass_Preferred
Formulation: Liquid. In SDS-PAGE sample buffer. Applications: WB. Shipping: Shipped on Dry Ice. Long Term Storage: -80°C. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-666-695