Learn More
PROTEINONE Hela Nuclear Extract for pre-mRNA splicing, 200 ug
Supplier:  PROTEINONE P00021
Encompass_Preferred
This HeLa cell nuclear extract is specifically recommended for: 1) in vitro splicing; 2) protein-DNA/RNA and protein-protein interactions; and 3) source of individual splicing factors. Although this extract also contains most transcription factors, it is, however, not recommended for in vitro transcription assays.
Catalog No. NC1791975