Learn More
AdipoGen Helenalin (1 mg)
SDP
Supplier:  AdipoGen AGCN20435M001
Encompass_Preferred
Chemical. CAS: 6754-13-8. Formula: C15H18O4. MW: 262.3. Isolated from Arnica chamissonis. Anticancer compound. NF-kappaB inhibitor. Apoptosis inducer. Potent anti-inflammatory agent. Telomerase inhibitor. Anti-trypanosomal and antiprotozoal compound Antibiotic. Shows anti-proliferative effects in 3T3-L1 preadipocytes. Autophagy inducer.
Catalog No. 50-205-3603