Learn More
Medchemexpress LLC Helichrysetin | 62014-87-3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN40585MG
Encompass_Preferred
Helichrysetin, isolated from the flower Helichrysum odoratissimum, acts as an ID2 (DNA binding inhibitor 2) inhibitor. It induces apoptosis and possesses anti-tumor and antioxidant activities.
  • Inhibits ID2, a DNA binding inhibitor.
  • Induces apoptosis in various cancer cell lines.
  • Exhibits anti-tumor activity by inhibiting cancer cell growth.
  • Possesses antioxidant activity.
  • Blocks cell cycle progression.
  • Reduces expression of proteins involved in cancer metabolism.
Catalog No. 50-004-19294