Learn More
ATCC Helicobacter pylori
Supplier:  ATCC 43504
Encompass
Helicobacter pylori strain NCTC 11637 is a whole-genome sequenced bacterial type strain that was isolated from the human gastric antrum. This strain can be used in enteric research or as a quality control strain.
Catalog No. 50-238-4118