Learn More
Oz Biosciences Helix-IN Transfection Reagent, 1 mL
SDP
Supplier:  Oz Biosciences HX11000
Encompass
Helix-IN™ DNA Transfection Reagent opens up new possibilities for addressing issues of classical transfection technologies.OZ Biosciences revolutionizes Polyfection with the design of Helix-IN™, a novel patented Cationic Hydroxylated Amphiphilic Multi-block Polymer (CHAMP™ Technology). This novel bi-functional co-polymer is biocompatible, ionizable, pH responsive and biodegradable.Cellular stress is reduced, viability preserved and efficiency enhanced.Broad Spectrum transfection Reagent for cell lines and hard-to-transfect cellsHigh Intracellular Protein Production while preserving viabilityHigh secreted Protein Production while minimizing cellular stressBiodegradable - Avoid secondary effectsCompatible with any culture medium
Catalog No. NC3072387