Learn More
Bisco Industries HELLERMANNTYTON ZIP TIE 1000PK
SDP
Supplier:  Bisco Industries T50R0HSM4
Encompass
Bundle diameter:1.97 INColor:Black (BK)Length:8 INMaterial:polyamide 6.6 heat stabilized (PA66HS)Standard:UL Listed (US and Canada), UL Recognized (US and Canada)Type:T50RWidth:0.18 IN
Catalog No. NC1519068