Learn More
ACROBiosystems Influenza B (Phuket/3073/2013) Hemagglutinin (HA) Protein, His Tag, Recombinant, 100 μg
Supplier:  ACROBiosystems HAEV52H4100UG
Encompass_Preferred
Recombinant Influenza B (Phuket/3073/2013, B/Yamagata lineage) hemagglutinin (HA) protein with a C-terminal His tag, supplied as 100 μg lyophilized material. Produced in HEK293 cells and formulated for stability; suitable for ELISA and antibody characterization.
  • Expressed in human 293 (HEK293) cells.
  • C-terminal polyhistidine (His) tag for detection or purification.
  • Amino acid range Asp 16-Ser 558 (Accession: EPI529345).
  • Calculated molecular weight 64.1 kDa; migrates 80-100 kDa due to glycosylation.
  • Purity greater than 90% by SDS-PAGE.
  • Lyophilized from 0.1 M sodium citrate, pH 5.5, with trehalose as protectant.
  • Store lyophilized at -20 °C or lower; avoid repeated freeze-thaw.
  • Validated in ELISA with a linear detection range of 0.5-63 ng/mL.
Catalog No. 50-235-7352