Learn More
ACROBiosystems Influenza A (A/Darwin/9/2021, H3N2) Hemagglutinin (HA) Protein, His tag - 100 μg
Supplier: ACROBiosystems HA2V52H6100UG
Recombinant hemagglutinin (HA) protein from Influenza A (H3N2), expressed in HEK293 cells with a C-terminal His tag. Supplied lyophilized for long-term storage; suitable for antibody binding assays, immunogen studies, and viral entry research.
- Recombinant hemagglutinin suitable for immunoassays and binding studies.
- Expressed in HEK293 cells to support native glycosylation.
- C-terminal polyhistidine tag for purification and detection.
- Purity greater than 90% as verified by SDS-PAGE and SEC-MALS.
- Glycosylated protein shows higher apparent molecular weight on SDS-PAGE.
- Lyophilized formulation in sodium citrate with trehalose for stability.
- Store lyophilized at -20°C to -70°C; reconstituted material should be stored at -70°C and handled sterilely.
- QC tested for binding to anti-HA antibodies over a wide linear range.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.