Learn More
ACROBiosystems Hemagglutinin HA 100ug
Supplier:  ACROBiosystems HA1V52H6100UG
Encompass_Preferred
Influenza A [A/Victoria/2570/2019] Hemagglutinin (HA) Protein His Tag (HA1-V52H6) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - Ala 532 (Accession # EPI_ISL_417210 GISAID).
Catalog No. 50-253-4901